missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANKLE2 Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA531750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIPA531750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA531750 Fournisseur Invitrogen™ Code fournisseur PA531750
Il en reste null

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Jurkat, Raji. Predicted reactivity: Human (99%), Mouse (84%), Rat (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a human guanine nucleotide exchange factor. It transduces signals from CRK by binding the SH3 domain of CRK, and activating several members of the Ras family of GTPases. This signaling cascade that may be involved in apoptosis, integrin-mediated signal transduction, and cell transformation. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined.

Spécifications

Antigen ANKLE2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ANKLE2
Gene Accession No. Q86XL3
Gene Alias 1110001J12Rik; Ab2-034; AI661024; Ankle2; ankyrin repeat and LEM domain containing 2; ankyrin repeat and LEM domain-containing protein 2; D5Ertd585e; Kiaa0692; LEM domain containing 7; LEM domain-containing protein 4; LEM4; LEMD7; liver regeneration-related protein LRRG057; MCPH16; RGD1310191
Gene Symbols ANKLE2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ANKLE2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23141
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats