missing translation for 'onlineSavingsMsg'
Learn More

Aniracetam

Catalog No. AAJ6166103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6166103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6166103 Supplier Thermo Scientific Chemicals Supplier No. J6166103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 72432-10-1 | C12H13NO3 | 219.24 g/mol

AMPA receptor potentiator. Slows the rate of ion-channel closing

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • AMPA receptor potentiator
  • Slows the rate of ion-channel closing

Chemical Identifiers

CAS 72432-10-1
Molecular Formula C12H13NO3
Molecular Weight (g/mol) 219.24
MDL Number MFCD00153767
InChI Key ZXNRTKGTQJPIJK-UHFFFAOYSA-N
Synonym aniracetam, draganon, sarpul, 1-4-methoxybenzoyl-2-pyrrolidinone, ampamet, 1-4-methoxybenzoyl pyrrolidin-2-one, 1-p-anisoyl-2-pyrrolidinone, memodrin, aniracetamun inn-latin, aniracetam usan:inn:jan
PubChem CID 2196
ChEBI CHEBI:47943
IUPAC Name 1-(4-methoxybenzoyl)pyrrolidin-2-one
SMILES COC1=CC=C(C=C1)C(=O)N2CCCC2=O

Specifications

Color White to Yellow
Quantity 1 g
Merck Index 14,662
Solubility Information Soluble in chloroform; Also soluble in ethanol; Insoluble in water
Formula Weight 219.24
Physical Form Powder
Chemical Name or Material Aniracetam
RTECSNumber UY5781900
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only