missing translation for 'onlineSavingsMsg'
Learn More

Aniline, 99+%, ACS reagent

Catalog No. AC423420025
Change view
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423420025 2.5 L Glass bottle
AC423420050 5 mL Glass bottle
AC423425000 500 mL Glass bottle
AC423420010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC423420025 Supplier Thermo Scientific Chemicals Supplier No. 423420025
Only null left

CAS: 62-53-3 | C6H7N | 93.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62-53-3
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00007629
InChI Key PAYRUJLWNCNPSJ-UHFFFAOYSA-N
Synonym benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam
PubChem CID 6115
ChEBI CHEBI:17296
IUPAC Name aniline
SMILES NC1=CC=CC=C1

Specifications

Melting Point -6.2°C
Color Colorless to White
Density 1.0213g/mL
Boiling Point 181.0°C to 185.0°C
Flash Point 76°C
Ignition Residue 0.005 % max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5845 to 1.5865
Linear Formula C6H5NH2
Quantity 2.5 L
Beilstein 12, 59
Fieser 06,21; 17,16
Specific Gravity 1.0213
Merck Index 15, 652
Solubility Information Solubility in water: 36g/L (20°C).
Viscosity 4.4 mPa.s (20°C)
Vapor Pressure 0.5mmHg at 20°C
Formula Weight 93.13
Percent Purity 99+%
Grade ACS Reagent
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Aniline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Toxic in contact with skin.
Toxic if swallowed.
Causes serious eye dam
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 200-539-3
Recommended Storage this product darkens to reddish-brown on storage

RUO – Research Use Only