missing translation for 'onlineSavingsMsg'
Learn More

Anhydrous Citric Acid, Granular, USP, 99.5-100.5%, Spectrum™ Chemical

Catalog No. 18600702
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600702 500 g Poly Bottle
18600703 2.5 kg Poly Bottle
18600704 12 kg Poly Pail
18600706 25 kg Fiber Drum
18600705 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18600702 Supplier Spectrum Chemical Mfg Cor Supplier No. CI133500GM
Only null left

CI133, 77-92-9, C6H8O7

Spectrum™ Chemical Anhydrous Citric Acid, Granular, USP belongs to a class of drugs known as urinary alkalinizers that are used to treat certain metabolic problems (acidosis) caused by kidney disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

CAS Max % 100%
Percent Purity 99.5 to 100.5%
Grade USP
Quantity 500 g
Ignition Residue 0.1%
Packaging Poly Bottle