missing translation for 'onlineSavingsMsg'
Learn More

Angiotensin I (human)

Catalog No. AAJ62102MB
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62102MB 25 mg
AAJ62102LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62102MB Supplier Thermo Scientific Chemicals Supplier No. J62102MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 484-42-4 | C62H89N17O14 | 1296.50 g/mol

Angiotensin I (human) stimulates the release of aldosterone, another hormone, from the adrenal cortex. It acts as a precursor to angiotensin II. It acts as endogenous peptide substrate for angiotensin converting enzyme (ACE), precursor to the vasoconstrictor peptide angiotensin II

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Angiotensin I (human) stimulates the release of aldosterone, another hormone, from the adrenal cortex. It acts as a precursor to angiotensin II. It acts as endogenous peptide substrate for angiotensin converting enzyme (ACE), precursor to the vasoconstrictor peptide angiotensin II

Solubility
Soluble to 1 mg/ml in water.

Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.

Chemical Identifiers

CAS 484-42-4
Molecular Formula C62H89N17O14
Molecular Weight (g/mol) 1296.50
MDL Number MFCD00133091
InChI Key ORWYRWWVDCYOMK-HBZPZAIKSA-N
Synonym angiotensin i, human angiotensin i, angiotensin i human, 5-ile-angiotensin i, angiotensin i rat, angiotensin i mouse, angiotensin 1 human, angiotensin i human, mouse, rat, pepsitensin, ile5-angiotensin i
PubChem CID 3081372
ChEBI CHEBI:2718
IUPAC Name (2S)-2-[(2S)-2-[(2S)-2-{[(2S)-1-[(2S)-2-[(2S,3S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-amino-3-carboxypropanamido]-5-[(diaminomethylidene)amino]pentanamido]-3-methylbutanamido]-3-(4-hydroxyphenyl)propanamido]-3-methylpentanamido]-3-(1H-imidazol-5-yl)propanoyl]pyrrolidin-2-yl]formamido}-3-phenylpropanamido]-3-(1H-imidazol-5-yl)propanamido]-4-methylpentanoic acid
SMILES CC[C@H](C)[C@H](NC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)[C@@H](NC(=O)[C@H](CCCN=C(N)N)NC(=O)[C@@H](N)CC(O)=O)C(C)C)C(=O)N[C@@H](CC1=CN=CN1)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CC1=CC=CC=C1)C(=O)N[C@@H](CC1=CN=CN1)C(=O)N[C@@H](CC(C)C)C(O)=O

Specifications

Color White
Quantity 25 mg
Solubility Information Soluble to 1mg/ml in water.
Formula Weight 1296.5
Physical Form Powder
Chemical Name or Material Angiotensin I (human)
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only