Learn More
Aneroid Dry Calorimeter
Catalog No. S41747
Change view
Click to view available options
Includes:
Instructions with graphical workups
Catalog No. Includes
S41747 Instructions with graphical workups
1 options
Catalog No. S41747 Supplier Customer Channels Group (Ccg) Supplier No. 15120
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Instructions with graphical workups
Five times more sensitive than traditional versions! Safe for students of all ages.
  • Consists of an aluminum cylinder, surrounded by a layer of Styrofoam™ insulation, enclosed in a durable plastic case
  • Thermometer (not included) can easily be inserted in top of calorimeter
  • Minimal heat loss; yields precise measurements of changes in temperature
  • Can be used with specific heat specimens up to 6.4cm in size (sold separately, see S41738)
Specifications
Product Type Aneroid Dry Student Calorimeter
For Use With (Application) Five times more sensitive than traditional versions, can be used with specific heat specimens up to 6.4cm in size
Includes Instructions with graphical workups