missing translation for 'onlineSavingsMsg'
Learn More

Anatomical Chart Series - Diseases, Disorders, and Processes

Numéro de catalogue. p-3562313
Change view
Click to view available options
Product Type:
Dangers of Smoking Anatomical Chart
Hip/Knee Inflammation Anatomical Chart
Infertility Anatomical Chart
Skin and Common Disorders Chart
Format:
Laminated
Unmounted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Product Type Format
S23598DL Infertility Anatomical Chart Laminated
S9865DU Dangers of Smoking Anatomical Chart Unmounted
S24217PL Skin and Common Disorders Chart Laminated
S23602DL Hip/Knee Inflammation Anatomical Chart Laminated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. S23598DL Fournisseur Customer Channels Group (Ccg) Code fournisseur 9740DLE
Il en reste null

Setting the highest standard of detail found in anatomical charts today!

Extraordinary, highly detailed charts ideal for school use.

Both the 11 x 14 in. desk size and the 20 x 26 in. poster size are available unmounted or laminated; laminated charts come with a special water-soluble marking pen as a teaching aid.

Spécifications

Product Type Infertility Anatomical Chart
Format Laminated
For Use With (Application) Extraordinary, highly detailed charts ideal for school use
Subject Science