missing translation for 'onlineSavingsMsg'
Learn More

Anastrozole 98.0+%, TCI America™

Catalog No. A2370100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2370100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2370100MG Supplier TCI America Supplier No. A2370100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 120511-73-1
Molecular Formula C17H19N5
Molecular Weight (g/mol) 293.374
MDL Number MFCD00866298
InChI Key YBBLVLTVTVSKRW-UHFFFAOYSA-N
Synonym anastrozole, arimidex, anastrazole, anastrozol, 2,2'-5-1h-1,2,4-triazol-1-yl methyl-1,3-phenylene bis 2-methylpropanenitrile, anastrole, asiolex, zeneca zd 1033, unii-2z07myw1az, arimidex tn
PubChem CID 2187
ChEBI CHEBI:2704
IUPAC Name 2-[3-(2-cyanopropan-2-yl)-5-(1,2,4-triazol-1-ylmethyl)phenyl]-2-methylpropanenitrile
SMILES CC(C)(C#N)C1=CC(=CC(=C1)CN2C=NC=N2)C(C)(C)C#N

Specifications

Melting Point 84°C
Color White
Quantity 100 mg
UN Number 3439
Formula Weight 293.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Anastrozole
RTECSNumber CZ1465000
TSCA No