missing translation for 'onlineSavingsMsg'
Learn More

Amyltriethylammonium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. A22745G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A22745G Supplier TCI America Supplier No. A22745G
Only null left

Chemical Identifiers

CAS 906478-91-9
Molecular Formula C13H26F6N2O4S2
Molecular Weight (g/mol) 452.471
InChI Key ALYCOCULEAWWJO-UHFFFAOYSA-N
Synonym Triethylpentylammonium Bis(trifluoromethanesulfonyl)imide
PubChem CID 53384372
IUPAC Name bis(trifluoromethylsulfonyl)azanide;triethyl(pentyl)azanium
SMILES CCCCC[N+](CC)(CC)CC.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 452.47
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Amyltriethylammonium Bis(trifluoromethanesulfonyl)imide