missing translation for 'onlineSavingsMsg'
Learn More

Amyl 4-(4-Ethoxyphenoxycarbonyl)phenyl Carbonate 95.0+%, TCI America™

Catalog No. A06081G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A06081G Supplier TCI America Supplier No. A06081G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33926-46-4
Molecular Formula C21H24O6
Molecular Weight (g/mol) 372.42
MDL Number MFCD00059434
InChI Key QVUSHDHMLLZBDM-UHFFFAOYSA-N
Synonym 4-(4-Ethoxyphenoxycarbonyl)phenyl Pentyl Carbonate
PubChem CID 578675
IUPAC Name 4-ethoxyphenyl 4-{[(pentyloxy)carbonyl]oxy}benzoate
SMILES CCCCCOC(=O)OC1=CC=C(C=C1)C(=O)OC1=CC=C(OCC)C=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 372.42
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Amyl 4-(4-Ethoxyphenoxycarbonyl)phenyl Carbonate