missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Amrinone, 98%
Description
Phosphodiesterase III inhibitor and inhibits platelet aggregation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 250 mg |
| Description | Yellow |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Phosphodiesterase III inhibitor and inhibits platelet aggregation |
| Formula Weight | 187.20g/mol |
| Melting Point | 294°C to 297°C (dec.) |
| Solubility | Insoluble in water or chloroform; Slightly soluble in acetic acid or DMF; Soluble in DMSO |
| Product Type | Signal Transduction Reagent |
| Form | Crystals |
| Assay | 98% |
RUO – Research Use Only