Learn More
Beckman Coulter AMPure XP Reagent, 450 mL, quantity of one
Supplier:  Beckman Coulter A63882
Encompass
The agencourt aMPure XP system is a highly efficient, easily automated PCR purification system that delivers superior quality DNa with no salt carryover. Requiring no centrifugation or filtration, agencourt aMPure XP can be easily used in manual and automated 96- or 384-well formats.
Numéro de catalogue. NC0110018
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.