missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq™ DNA Polymerase, LD (Low DNA) DFS Item

Applied Biosystems AmpliTaq DNA Polymerase, LD (Low DNA), is the same enzyme as AmpliTaq DNA Polymerase, but is further purified through a proprietary separation process to ensure that residual bacterial DNA sequences are substantially reduced.

Supplier:  Applied Biosystems™ N8080158

 View more versions of this product

Catalog No. N8080158

Only null left
Explore available promotions

Includes: Contains 250 units of AmpliTaq DNA Polymerase, Buffer II, and MgCl2 Solution.

Description

Description

Applied Biosystems AmpliTaq DNA Polymerase, LD (Low DNA), is the same enzyme as AmpliTaq DNA Polymerase, but is further purified through a proprietary separation process to ensure that residual bacterial DNA sequences are substantially reduced.

This highly purified enzyme preparation ensures that non-target, false-positive PCR products will be effectively minimized when amplifying bacterial sequences. This is especially useful for low-copy-number PCR amplifications. The enzyme is quality-control tested to verify that fewer than 10 copies of bacterial 16S ribosomal RNA gene sequences are present in a standard 2.5 unit aliquot.

AmpliTaq DNA Polymerase, LD, is supplied with either 10X PCR Buffer I or with 10X PCR Buffer II and MgCl2 Solution.

Features

  • Minimizes non-specific PCR products when amplifying bacterial targets
  • Highly purified enzyme making it particularly useful for low copy number amplifications

Notes

  • See user's manual or package insert for limited label license and trademark information.

Order Info

Shipping Condition: Dry ice

Specifications

Specifications

5 U/μL
Stand-alone enzyme
AmpliTaq DNA Polymerase
5' - 3'
LD (Low DNA)
Dry Ice
1X
200 Reactions
Separate Components
DNA
• AmpliTaq DNA Polymerase, LD (5 U/μL), 25 μL
• 10X PCR Buffer II, 1.5 mL
• 25 mM MgCl2 Solution, 1.5 mL

Store at -15°C to -30°C.
High
Standard
DNA Polymerase
250 units
Standard PCR
No
3'-A
5 kb or less
Buffer II
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.