missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq™ DNA Polymerase, LD (Low DNA)

Applied Biosystems AmpliTaq DNA Polymerase, LD (Low DNA), is the same enzyme as AmpliTaq DNA Polymerase, but is further purified through a proprietary separation process to ensure that residual bacterial DNA sequences are substantially reduced.

$403.00 - $417.00

Specifications

Concentration 5 U/μL
Format Stand-alone enzyme
GC-Rich PCR Performance High
Polymerase AmpliTaq DNA Polymerase
Reaction Speed Standard
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
N8080157
Encompass
View Documents Promotion Details
Applied Biosystems™
N8080157
Buffer I
Each for $417.00
Only null left
 
N8080158
Encompass
View Documents Promotion Details
Applied Biosystems™
N8080158
Buffer II
Each for $403.00
Only null left
 
Description

Description

Applied Biosystems AmpliTaq DNA Polymerase, LD (Low DNA), is the same enzyme as AmpliTaq DNA Polymerase, but is further purified through a proprietary separation process to ensure that residual bacterial DNA sequences are substantially reduced.

This highly purified enzyme preparation ensures that non-target, false-positive PCR products will be effectively minimized when amplifying bacterial sequences. This is especially useful for low-copy-number PCR amplifications. The enzyme is quality-control tested to verify that fewer than 10 copies of bacterial 16S ribosomal RNA gene sequences are present in a standard 2.5 unit aliquot.

AmpliTaq DNA Polymerase, LD, is supplied with either 10X PCR Buffer I or with 10X PCR Buffer II and MgCl2 Solution.

Features

  • Minimizes non-specific PCR products when amplifying bacterial targets
  • Highly purified enzyme making it particularly useful for low copy number amplifications

Notes

  • See user's manual or package insert for limited label license and trademark information.
Specifications

Specifications

5 U/μL
High
Standard
DNA Polymerase
250 units
Standard PCR
No
3'-A
5 kb or less
Stand-alone enzyme
AmpliTaq DNA Polymerase
5' - 3'
LD (Low DNA)
Dry Ice
1X
200 Reactions
Separate Components
DNA
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.