missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin Sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015262
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015262 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015262 Supplier Merck Emd Millipore Supplier No. PHR14241G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 69-52-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C16H18N3 NaO4 S
MDL Number MFCD00064313
Melting Point 215°C
Synonym Ampicillin sodium salt; D-(-)-alpha-Aminobenzylpenicillin sodium salt
Type Certified Reference Material
Quantity 1 g
Formula Weight 371.39
Grade Certified Reference Material