missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin, ≥95% anhydrous basis (NT), MilliporeSigma™ Supelco™

Catalog No. 111015263
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015263 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015263 Supplier Merck Emd Millipore Supplier No. 59349100MG

Chemical structure: β-lactamAmpicillin belongs to the β-lactum group of antibiotics. It is used to treat bacterial infections via penetrating gram positive and gram negative bacteria. Its mode of action basically involves the inhibition of bacterial cell wall synthesis, thus leading to cell lysis.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 69-53-4
Physical Form Neat
Linear Formula C16H19N3O4 S
MDL Number MFCD00005175
Melting Point 208°C
Synonym D-(-)-alpha-Aminobenzylpenicillin
Type Analytical Standard
Quantity 100 mg
Formula Weight 349.40
Percent Purity ≥95% anhydrous basis (NT)
Grade Analytical Standard
Show More Show Less