missing translation for 'onlineSavingsMsg'
Learn More

AMPD, 0.2M buffer soln., pH 9.5, Thermo Scientific™

Catalog No. AAJ60916AK
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60916AK 250 mL
AAJ60916AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60916AK Supplier Thermo Scientific Chemicals Supplier No. J60916AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 115-69-5
Color Colorless
Concentration 0.2 M
pH 9.5
Physical Form Liquid
Quantity 250 mL
Odor Odorless
Molecular Weight (g/mol) 105.137