missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tungsten oxide hydrate

Catalog No. AA4479236
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4479236 500 g
AA4479214 25 g
AA4479222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4479236 Supplier Thermo Scientific Chemicals Supplier No. 04479236
Only null left

CAS: 12333-11-8 | (NH4)6W12O39·xH2O

Ammonium tungsten oxide hydrate is widely utilized as a raw material for the synthesis of tungsten catalysts, which is used in a variety of reactions like oxidation, hydroxylation, hydrogenation and polymerization. It is used as a reagent for chemical analysis like medical diagnosis. It is used as a precursor material for the preparation of high-purity WO3 which is used as thin-film substrate for certain semiconductor devices. It acts as a precursor to a variety of POM (polyoxometalates).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12333-11-8
Molecular Formula (NH4)6W12O39·xH2O
MDL Number MFCD00042673
Synonym Ammonium metatungstate hydrate

Specifications

Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 2938.41 (Anhydrous)
Chemical Name or Material Ammonium tungsten oxide hydrate
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 683-059-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only