missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, ACS Grade, ≥ 97.5%, LabChem™

Catalog No. LC113401
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC113401 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC113401 Supplier LabChem Supplier No. LC113401

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Melting Point 150°C
Color White
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula NH4SCN
Flash Point 190°C
Solubility Information Soluble in water
Formula Weight 76.12
Percent Purity ≥97.5%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.3g/cm3
Decomposition Information Nitrogen oxides; Carbon dioxide; Carbon monoxide; Sulfur compounds; Gaseous Ammonia
Density 1.3g/cm3
Chemical Name or Material Ammonium Thiocyanate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed, in contact with skin or if inhaled.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid breathing dust.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 217-175-6
Recommended Storage Room Temperature