Learn More
Ammonium thiocyanate, ACS, 97.5% min
Description
Ammonium thiocyanate has many uses in manufacturing chemicals such as metal thiocyanates [CuSCN, Ca(SCN)2, NaSCN, KSCN], and in a number of pharmaceuticals. It is used in freezing solutions, pickling, printing, and corrosion inhibitor against acid gases. It is used in photography industry as a stabilizer or accelerator. Used in fertilizers, fabric dyeing, zinc coating, electroplating, and as a separator of Zr and Hf, also Au and Fe.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 147°C to 149°C |
| Physical Form | Crystalline |
| Quantity | 1 kg |
| Sensitivity | Hygroscopic; Light sensitive |
| Assay Percent Range | 97.5% min |
| Density | 1.305 |
| Beilstein | 3595135 |
| Solubility Information | Soluble in water,ethanol,methanol,ammonia,and acetone. Practically insoluble in CHCl3and ethyl acetate. |
| Formula Weight | 76.12 |
| Grade | ACS |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332-H318 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c |
| EINECSNumber | 217-175-6 |
| RTECSNumber | XK7875000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only