missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, 30% (w/v), Ricca Chemical

Catalog No. R069800020F
Change view
Click to view available options
Package: Cubitainer; Size: 20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
R069800020F Package: Cubitainer; Size: 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R069800020F Supplier Ricca Chemical Company Supplier No. R069800020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 1762-95-4, 7732-18-5
CAS Min % 75.0, 29.0
CAS Max % 78.0, 31.0
Molecular Formula CH4N2S
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
SMILES C(#N)[S-].[NH4+]
IUPAC Name azanium;thiocyanate
Molecular Weight (g/mol) 76.117
PubChem CID 15666
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
Quantity 20 L
Concentration or Composition Notes 30% (w/v)
Chemical Name or Material Ammonium Thiocyanate
Show More Show Less