missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Thiocyanate, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 1 L |
| Formula Weight | 76.12 |
| Grade | BAKER ANALYZED™ Reagent |
| Packaging | Narrow Mouth Polyethylene Bottle |
| Specific Gravity | 1L = 1.10kg |
| Chemical Name or Material | Ammonium Thiocyanate, 0.1N Volumetric Solution |