missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, 0.0250 N (N/40), Ricca Chemical

Catalog No. p-3251312
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
694716 500 mL Natural Poly Bottle
694732 1 L Natural Poly Bottle
69471 4 L Natural Poly Bottle
R069470020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 694716 Supplier Ricca Chemical Company Supplier No. 694.716

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1762-95-4 , 7732-18-5
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

CAS Min % 98.0, 0.19
CAS Max % 100.0, 0.19
Packaging Natural Poly Bottle
Quantity 500 mL
Concentration or Composition Notes 0.025Normal
Chemical Name or Material Ammonium Thiocyanate