missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfide TS, Ricca Chemical

Catalog No. R0694600120
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0694600120 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R0694600120 Supplier Ricca Chemical Company Supplier No. R0694600120C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7783-08-6 , 1336-21-6 , 7783-06-4
Molecular Formula K2O4Se
Molecular Weight (g/mol) 221.16
InChI Key YAZJAPBTUDGMKO-UHFFFAOYSA-L
Synonym selenic vi acid, h2seo4, unii-hv0y51nc4j, hv0y51nc4j, hsdb 675, dihydroxidodioxidoselenium, h2so4e, aq. soln., seo2 oh 2
PubChem CID 1089
ChEBI CHEBI:18170
IUPAC Name selenic acid
SMILES O[Se](=O)(=O)O

Specifications

Color Yellow
CAS Min % 64.0, 23.4, 10.4
CAS Max % 67.0, 24.36, 10.82
Grade Laboratory
Packaging Amber Glass Bottle
pH 9.5
Quantity 120 mL
Physical Form Liquid
Chemical Name or Material Ammonium Sulfide, Test Solution