accessibility menu, dialog, popup

UserName

KKOSeOOO

CAS RN 7783-08-6

IUPAC Name: dipotassium selenate
Synonyms: potassium selenate
Molecular Weight (g/mol): 221.16
Molecular Formula: K2O4Se
InChi Key: YAZJAPBTUDGMKO-UHFFFAOYSA-L
SMILES: [K+].[K+].[O-][Se]([O-])(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
pH 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)