missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfide Solution, Reagent, ACS, 20-24%, Spectrum™ Chemical
Description
Spectrum™ Chemical Ammonium Sulfide Solution, Reagent, ACS is an unstable salt with the formula (NH4)2S. Common uses of AMMONIUM SULFIDE are industrial applications including photo developing and textile manufacturing. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Specifications
Specifications
| CAS Max % | 80%, 20% |
| Quantity | 20 L |
| UN Number | UN2683 |
| Percent Purity | 20 to 24% |
| Grade | ACS |
| Packaging | Steel Can |
| Residue after Ignition | 0.04% |