missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, 50% Saturated, with Hydrochloric Acid, Ricca Chemical

Catalog No. 694116
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
694116 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 694116 Supplier Ricca Chemical Company Supplier No. 694.116
Only null left

Precipitating Solution for Fetal Hemoglobin

Chemical Identifiers

CAS 7783-20-2 , 7647-01-0 , 7732-18-5
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Grade Laboratory
Packaging Natural Poly Bottle
Quantity 500 mL
Chemical Name or Material Ammonium Sulfate, 50% Saturated, with Hydrochloric Acid
ShelfLife 12 months