missing translation for 'onlineSavingsMsg'
Learn More

Ammonium phosphate monobasic, Analytical Standard, 99.5%, MilliporeSigma™ Supelco™

Catalog No. 111015492
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015492 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015492 Supplier Merck Emd Millipore Supplier No. 8843125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Nitrogen Determination according to Kjeldahl Method

Specifications

CAS 7722-76-1
Physical Form Neat
Linear Formula NH4H2PO4
MDL Number MFCD00003396
Melting Point 190°C (decomposition)
Synonym mono-Ammonium phosphate; prim-Ammonium phosphate; Ammonium dihydrogenphosphate
Solubility Soluble in water
Quantity 25 g
Formula Weight 115.03
Percent Purity ≥99.5% (T)
Grade Analytical Standard
Show More Show Less