missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Dibasic, 30% (w/v), Ricca Chemical

Catalog No. p-3340183
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6893016 500 mL Natural Poly Bottle
6893032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6893016 Supplier Ricca Chemical Company Supplier No. 689.3016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diammonium Hydrogen Phosphate Solution, APHA for Magnesium

Chemical Identifiers

CAS 7783-28-0 , 7732-18-5
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.056
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
IUPAC Name diazanium;hydrogen phosphate
SMILES [NH4+].[NH4+].OP(=O)([O-])[O-]

Specifications

CAS Min % 69.0, 29.0
CAS Max % 71.0, 31.0
Quantity 500 mL
Concentration or Composition (by Analyte or Components) 30% (w/v)
Odor Odorless
Packaging Natural Poly Bottle
Chemical Name or Material Ammonium Phosphate, Dibasic
ShelfLife 24 months