missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, Molecular Biology Reagent, MP Biomedicals™

Catalog No. ICN19398825
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19398825 25 g
ICN19398890 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19398825 Supplier MP Biomedicals Inc Supplier No. 0219398825

Ammonium Persulfate, Molecular Biology Reagent is a polymerization catalyst used with TEMED for polyacrylamide gel formation.

  • Ammonium persulfate is a polymerization catalyst used with TEMED for polyacrylamide gel formation.
  • APS has also been utilized to study protein-protein interactions via photoinitiated crosslinking chemistry.
  • Other applications of APS include its use as a reducer and retarder in photography, the manufacture of aniline dyes, electroplating, and the decolorizing and deodorizing of oils.

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C (Decomposition)
Color White
Physical Form Crystalline Powder
Quantity 25 g
Solubility Information Soluble in water, 83.5 g/100 g water at 25°C
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Formula Weight 228.18
Percent Purity ≥98%
Grade Molecular Biology
Density 1.98 g/cm3 at 68°F; Bulk Density: 951 to 1050 kg/m3
Chemical Name or Material Ammonium persulfate
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.