missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Peroxydisulfate, Crystal, ACS, 98%, Spectrum™ Chemical

Catalog No. 18608748
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
25 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608748 50 kg Poly Bottle
18608743 25 g Poly Bottle
18608744 100 g Poly Bottle
18608745 500 g Poly Bottle
18608746 2.5 kg Poly Bottle
18608747 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18608748 Supplier Spectrum Chemical Mfg Cor Supplier No. A122550KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle
Residue after Ignition 0.0005
Chemical Name or Material Ammonium Peroxydisulfate, Crystal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.