missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Peroxodisulfate 99.0+%, TCI America™

Catalog No. A20985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20985G 5 g
A209825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20985G Supplier TCI America Supplier No. A20985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
UN Number 1444
Formula Weight 228.19
Percent Purity ≥99.0% (T)
Grade Electrophoresis
Chemical Name or Material Ammonium Peroxodisulfate [for Electrophoresis]
EINECSNumber (1)-0406
RTECSNumber SE0350000
TSCA Yes