missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Oxalate TS, 3.5% (w/v), Ricca Chemical

Catalog No. 6791
Change view
Click to view available options
Poly natural; 4L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
6791 Poly natural; 4L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6791 Supplier Ricca Chemical Company Supplier No. RI6791

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Meets USP TS and ACS RS specifications

Specifications

CAS 6009-70-7, 7732-18-5
CAS Min % 95.0, 3.32
CAS Max % 99.0, 3.45
Molecular Formula C2H10N2O5
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O
IUPAC Name diazanium;oxalate;hydrate
Molecular Weight (g/mol) 142.111
PubChem CID 516808
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 4 L
Concentration or Composition Notes 3.5% (w/v)
Chemical Name or Material Ammonium Oxalate, Test Solution
Show More Show Less