missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Oxalate, Saturated, Ricca Chemical

Catalog No. R06840001A
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R06840001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R06840001A Supplier Ricca Chemical Company Supplier No. R06840001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6009-70-7 , 7732-18-5
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.111
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
PubChem CID 516808
IUPAC Name diazanium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Specifications

CAS Min % 93.0, 4.67
CAS Max % 97.0, 4.86
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 1 L
Chemical Name or Material Ammonium Oxalate, Saturated