missing translation for 'onlineSavingsMsg'
Learn More

Ammonium oxalate monohydrate, 99.5%, for analysis

Catalog No. p-59645
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC206275000 500 g Plastic bottle
AC206270010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC206275000 Supplier Thermo Scientific Chemicals Supplier No. 206275000

CAS: 6009-70-7 | C2H8N2O4·H2O | 142.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6009-70-7
Molecular Formula C2H8N2O4·H2O
Molecular Weight (g/mol) 142.11
MDL Number MFCD00149694
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
PubChem CID 516808
IUPAC Name diazanium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Specifications

Melting Point 70.0°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99.50%
Linear Formula (CO2NH4)2·H2O
Merck Index 15, 534
Solubility Information Solubility in water: 45g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 142.11
Percent Purity 99.5%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Ammonium oxalate monohydrate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only