missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Oxalate, For Calcium, Certified, 4.0% (w/v) ±0.1% (w/v), LabChem™
Supplier: LabChem 112802
Specifications
| Ammonium Oxalate | |
| For Calcium | |
| 96,4 | |
| C2H8N2O4·H2O | |
| ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate | |
| MSMNVXKYCPHLLN-UHFFFAOYSA-N | |
| diazanium;oxalate;hydrate | |
| 516808 | |
| 4.0% ±0.1% (w/v) | |
| Passes Test | |
| 1.04g/mL | |
| Gaseous Ammonia; Carbon monoxide; Carbon dioxide | |
| Liquid |
| Colorless | |
| 6009-70-7 , 7732-18-5 | |
| C2H10N2O5 | |
| 1.04g/mL | |
| Soluble in water | |
| C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O | |
| 142.111 | |
| 142.11 | |
| Certified | |
| Poly Bottle | |
| Traceable to NIST | |
| 1 L |
Chemical Identifiers
| 6009-70-7 | |
| 142.111 | |
| ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate | |
| diazanium;oxalate;hydrate |
| C2H10N2O5 | |
| MSMNVXKYCPHLLN-UHFFFAOYSA-N | |
| 516808 | |
| C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature