missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Oxalate, For Calcium, Certified, 4.0% (w/v) ±0.1% (w/v), LabChem™

Catalog No. p-5388360
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC112801 500 mL
LC112802 1 L
LC112804 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC112801 Supplier LabChem Supplier No. LC112801
Only null left

Chemical Identifiers

CAS 6009-70-7 , 7732-18-5
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.111
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
PubChem CID 516808
IUPAC Name diazanium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Specifications

Color Colorless
CAS Max % 96, 4
Linear Formula C2H8N2O4·H2O
Density 1.04g/mL
Solubility Information Soluble in water
Formula Weight 142.11
Percent Purity 4.0% ±0.1% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.04g/mL
Traceability to NIST Traceable to NIST
Decomposition Information Gaseous Ammonia; Carbon monoxide; Carbon dioxide
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Ammonium Oxalate, For Calcium
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature