missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Oxalate, 3.5% (w/v), LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 96.5, 3.5 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Linear Formula | C2H8N2O4·H2O |
| Solubility Information | Soluble in water |
| Formula Weight | 142.11 |
| Percent Purity | 3.5% (w/v) |
| Grade | Analytical |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |