missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate-Vanadate Solution, Ricca Chemical

Catalog No. 67732
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67732 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67732 Supplier Ricca Chemical Company Supplier No. 67732

For Phosphorus Determination

Specifications

Chemical Name or Material Ammonium Molybdate-Vanadate Solution
Color Yellow
CAS 7732-18-5, 7697-37-2, 12054-85-2, 7803-55-6
CAS Min % 73.0, 21.95, 2.92, 0.07
CAS Max % 76.0, 22.85, 3.04, 0.08
Synonym dihydrogen oxide, dihydrogen monoxide
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 1 L
Physical Form Liquid