missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate Solution, Saturated, Ricca Chemical Company

Catalog No. R06715001A
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R06715001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R06715001A Supplier Ricca Chemical Company Supplier No. R06715001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12054-85-2 , 7732-18-5

Specifications

Color Colorless to Light Green
Boiling Point ~100°C
Grade Laboratory
Quantity 1 L
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Ammonium Molybdate Solution
ShelfLife 18 Months