missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Molybdate Reagent II, For Phosphorus, LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 5.08, 77.86, 17.06 |
| Linear Formula | (NH4)6Mo7O24·4H2O |
| UN Number | UN2796 |
| Density | 1.1g/mL |
| Solubility Information | Soluble in water |
| Formula Weight | 1235.86 |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Specific Gravity | 1.1g/mL |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Wear protective gloves/eye protection. Wash exposed skin thoroughly after handling. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |