missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate Reagent I, For Phosphorus, Certified, LabChem™

Supplier:  LabChem LC112302

 View more versions of this product

Catalog No. LC112302


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Molybdate Reagent I
For Phosphorus
2.5,57.98,39.52
(NH4)6Mo7O24·4H2O
1.3g/mL
Soluble in water
[NH4+].O.[O--].[Mo]
148.00
1235.86
Passes Test
1.3g/mL
1.9cSt
1 L
Colorless
12054-85-2 , 7732-18-5 , 7664-93-9
H6MoNO2
UN2796
Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate
XCOMAVCHADVANQ-UHFFFAOYSA-O
ammonium hydrate molybdenum oxidandiide
3084102
Certified
Poly Bottle
Passes Test
Sulfur compounds; Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Liquid

Chemical Identifiers

12054-85-2
148.00
Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate
ammonium hydrate molybdenum oxidandiide
H6MoNO2
XCOMAVCHADVANQ-UHFFFAOYSA-O
3084102
[NH4+].O.[O--].[Mo]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions