missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate Reagent I, For Phosphorus, Certified, LabChem™

Catalog No. p-5388354
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC112301 500 mL
LC112302 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC112301 Supplier LabChem Supplier No. LC112301
Only null left

Chemical Identifiers

CAS 12054-85-2 , 7732-18-5 , 7664-93-9
Molecular Formula H6MoNO2
Molecular Weight (g/mol) 148.00
InChI Key XCOMAVCHADVANQ-UHFFFAOYSA-O
Synonym Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate
PubChem CID 3084102  
IUPAC Name ammonium hydrate molybdenum oxidandiide
SMILES [NH4+].O.[O--].[Mo]

Specifications

Color Colorless
CAS Max % 2.5, 57.98, 39.52
Linear Formula (NH4)6Mo7O24·4H2O
UN Number UN2796
Density 1.3g/mL
Solubility Information Soluble in water
Formula Weight 1235.86
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.3g/mL
Suitability Passes Test
Viscosity 1.9cSt
Decomposition Information Sulfur compounds; Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Ammonium Molybdate Reagent I, For Phosphorus
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature