missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate, For Silica, Certified, 10% (w/v), LabChem™

Catalog No. LC112501
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC112501 500 mL
LC112502 1 L
LC112504 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC112501 Supplier LabChem Supplier No. LC112501

Chemical Identifiers

CAS 12054-85-2 , 7732-18-5 , 1310-73-2
Molecular Formula H6MoNO2
Molecular Weight (g/mol) 148.00
InChI Key XCOMAVCHADVANQ-UHFFFAOYSA-O
Synonym Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate
PubChem CID 3084102  
IUPAC Name ammonium hydrate molybdenum oxidandiide
SMILES [NH4+].O.[O--].[Mo]

Specifications

Color Colorless
CAS Max % 9.69, 87.91, 2.4
Linear Formula (NH4)6Mo7O24·4H2O
Density 1.02g/mL
Solubility Information Soluble in water
Formula Weight 1235.86
Percent Purity 10% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.02g/mL
Suitability Passes Test
Viscosity 1.1cSt
Decomposition Information Gaseous Ammonia
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Ammonium Molybdate, For Silica
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature