missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate, 99.99%, (trace metal basis)

Catalog No. AC448540050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC448540050 5 g Glass bottle
AC448540250 25 g Glass bottle
AC448541000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC448540050 Supplier Thermo Scientific Chemicals Supplier No. 448540050

CAS: 13106-76-8 | H8MoN2O4 | 196.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13106-76-8
Molecular Formula H8MoN2O4
Molecular Weight (g/mol) 196.01

Specifications

Quantity 5 g
Assay Percent Range 99.99%
Linear Formula (NH4)2MoO4
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Formula Weight 196.01
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ammonium molybdate
Hazard Category GHS Signal Word: Warning
EINECSNumber 236-031-3

RUO – Research Use Only