missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Molybdate, 7.5% (w/v), pH adjusted, Ricca Chemical
$243.40 - $243.40
Chemical Identifiers
| CAS | 12054-85-2 |
|---|---|
| Molecular Formula | H6MoNO2 |
| Molecular Weight (g/mol) | 148.00 |
| InChI Key | XCOMAVCHADVANQ-UHFFFAOYSA-O |
| Synonym | Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate |
| PubChem CID | 3084102 |
| IUPAC Name | ammonium hydrate molybdenum oxidandiide |
| SMILES | [NH4+].O.[O--].[Mo] |
Chemical Identifiers
| 12054-85-2 | |
| 148.00 | |
| Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate | |
| ammonium hydrate molybdenum oxidandiide |
| H6MoNO2 | |
| XCOMAVCHADVANQ-UHFFFAOYSA-O | |
| 3084102 | |
| [NH4+].O.[O--].[Mo] |
Specifications
| Colorless | |
| 7.0,88.0,0.1 | |
| H6MoNO2 | |
| XCOMAVCHADVANQ-UHFFFAOYSA-O | |
| ammonium hydrate molybdenum oxidandiide | |
| 3084102 | |
| Laboratory | |
| pH adjusted | |
| 7 to 8 | |
| Liquid | |
| Ammonium Molybdate |
| 12054-85-2 , 7732-18-5 , 1310-73-2 | |
| 8.0,92.0,0.9 | |
| Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate | |
| [NH4+].O.[O--].[Mo] | |
| 148.00 | |
| 7.5% | |
| Natural Poly Bottle | |
| Aqueous Solution | |
| 500 mL | |
| 7.5% (w/v) |