missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate, 10% (w/v), pH adjusted, Ricca Chemical

Catalog No. p-3251297
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
67016 500 mL Natural Poly Bottle
67032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 67016 Supplier Ricca Chemical Company Supplier No. 67016
Only null left

Chemical Identifiers

CAS 12054-85-2 , 7732-18-5 , 1310-73-2
Molecular Formula H6MoNO2
Molecular Weight (g/mol) 148.00
InChI Key XCOMAVCHADVANQ-UHFFFAOYSA-O
Synonym Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate
PubChem CID 3084102
IUPAC Name ammonium hydrate molybdenum oxidandiide
SMILES [NH4+].O.[O--].[Mo]

Specifications

Color Colorless
CAS Min % 9.0, 85.0, 0.1
CAS Max % 11.0, 88.0, 0.9
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
Treatment(s) pH adjusted
Solvent or Matrix Aqueous Solution
pH 7 to 8
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 10% (w/v)
Chemical Name or Material Ammonium Molybdate
Show More Show Less