missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium iron(III) sulfate dodecahydrate, 99%, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 230.0°C |
| Melting Point | 39.0°C to 41.0°C |
| Color | Violet |
| Physical Form | Crystals |
| Quantity | 2.5 kg |
| Assay Percent Range | 98.5 to 102% (Iodometry) |
| Linear Formula | (NH4)Fe(SO4)2·12H2O |
| Merck Index | 15, 515 |
| Solubility Information | Solubility in water: 1240g/L water (25°C). Other solubilities: practically insoluble in alcohol |
| Formula Weight | 482.19 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. Wear protective gloves/protective clothing/eye protec |
| RTECSNumber | WS5900000 |
RUO – Research Use Only