missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iron(Ii) Sulfate Hexahydrate, puriss. p.a., Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 1 kg |
| Linear Formula | (NH4)2Fe(SO4)2 · 6H2O |
| Formula Weight | 392.14g/mol |
| Percent Purity | ≥99% (RT) |
| Grade | puriss. p.a. |
| Chemical Name or Material | Ammonium Iron(Ii) Sulfate Hexahydrate, Reag. Ph. Eur. reag. ISO |
Safety and Handling
| EINECSNumber | 233-151-8 |
| RTECSNumber | BR6500000 |