missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, Ultratrace, 20-22%, Spectrum™ Chemical

Catalog No. 18608686
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608686 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18608686 Supplier Spectrum Chemical Mfg Cor Supplier No. A1076500MLPL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1336-21-6 , 7664-41-7
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
IUPAC Name amine hydrate
SMILES N.O

Specifications

CAS Max % 1
Quantity 500 mL
Percent Purity 20 to 22%
Grade High Purity Solvent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.